Request a Quote - ÄKTA
Cytiva
ÄKTA™ start protein purification system
ÄKTA start offers the following features:
  • Compact solution for quick and reliable one-step protein purification
  • Sample injection and fraction collection options
  • Quick start methods and templates for common purification techniques
  • Intuitive touchscreen display and real-time monitoring
Specifications:
  • Flow rate: 0.5 to 5mL/min
  • Pump type: peristaltic pump - single channel, four-roller pump head with low pulsation
  • Max. operating pressure: 0.5MPa
  • UV, fixed wavelength: Yes, 280nm
  • Conductivity range: 0 to 300mS/cm
  • Software: UNICORN start 1.0 or higher