man

Test

CAS RN 10026-17-2

FFCo

CAS RN 10026-17-2

IUPAC Name: λ²-cobalt(2+) difluoride
Synonyms: cobalt(II) fluoride
Molecular Weight (g/mol): 96.93
Molecular Formula: CoF2
InChi Key: YCYBZKSMUPTWEE-UHFFFAOYSA-L
SMILES: [F-].[F-].[Co++]
Molecular Weight (g/mol) 
  • (5)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (5)
  • (2)
  • (2)
  • (2)
  • (3)
  • (7)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)