accessibility menu, dialog, popup

Test

CAS RN 1003-29-8

ONH

CAS RN 1003-29-8

IUPAC Name: 1H-pyrrole-2-carbaldehyde
Synonyms: pyrrole-2-carboxaldehyde 2-carboxaldehyde-1H-pyrrole 1(H)-pyrrole carboxaldehyde 2-pyrrolylcarboxaldehyde 1-pyrrole-2-carboxaldehyde
Molecular Weight (g/mol): 95.1
Molecular Formula: C5H5NO
InChi Key: ZSKGQVFRTSEPJT-UHFFFAOYSA-N
SMILES: O=CC1=CC=CN1
Molecular Weight (g/mol) 
  • (3)
  • (4)
Percent Purity 
  • (1)
  • (6)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
Boiling Point 
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (3)
  • (4)
  • (1)
  • (6)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (3)