accessibility menu, dialog, popup

Test

CAS RN 1003-61-8

CAS RN 1003-61-8

IUPAC Name: 2-amino-1,3-thiazole-5-carbaldehyde
Synonyms: 2-amino-1,3-thiazole-5-carbaldehyde
Molecular Weight (g/mol): 128.15
Molecular Formula: C4H4N2OS
InChi Key: HZKMBJCDAXLMDN-UHFFFAOYSA-N
SMILES: NC1=NC=C(S1)C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results