CAS RN 1005775-56-3
CAS RN 1005775-56-3
IUPAC Name:
4-tert-butyl-N-[6-(1H-imidazol-1-yl)imidazo[1,2-a]pyridin-2-yl]benzamide dihydrochloride
Synonyms:
4-tert-butyl-N-[6-(imidazol-1-yl)imidazo[1,2-a]pyridin-2-yl]benzamide dihydrochloride
Molecular Weight (g/mol):
432.35
Molecular Formula:
C21H23Cl2N5O
InChi Key:
IKKLFEDUYFZNBO-UHFFFAOYSA-N
SMILES:
Cl.Cl.CC(C)(C)C1=CC=C(C=C1)C(=O)NC1=CN2C=C(C=CC2=N1)N1C=CN=C1
1
–
1
of
1
results
1
TC ASK 10, Tocris Bioscience™
CAS: 1005775-56-3 Molecular Formula: C21H23Cl2N5O Molecular Weight (g/mol): 432.349 InChI Key: IKKLFEDUYFZNBO-UHFFFAOYSA-N Synonym: ask1 inhibitor 10,tc ask 10,4-tert-butyl-n-6-1h-imidazol-1-yl imidazo 1,2-a pyridin-2-yl benzamide dihydrochloride,4-1,1-dimethylethyl-n-6-1h-imidazol-1-yl imidazo 1,2-a pyridin-2-yl benzamide dihydrochloride PubChem CID: 68661090 IUPAC Name: 4-tert-butyl-N-(6-imidazol-1-ylimidazo[1,2-a]pyridin-2-yl)benzamide;dihydrochloride SMILES: CC(C)(C)C1=CC=C(C=C1)C(=O)NC2=CN3C=C(C=CC3=N2)N4C=CN=C4.Cl.Cl