CAS RN 1007-15-4
CAS RN 1007-15-4
IUPAC Name:
1-(3-bromo-4-fluorophenyl)ethan-1-one
Synonyms:
1-(3-bromo-4-fluorophenyl)ethanone
Molecular Weight (g/mol):
217.04
Molecular Formula:
C8H6BrFO
InChi Key:
SZDWTGAORQQQGY-UHFFFAOYSA-N
SMILES:
CC(=O)C1=CC=C(F)C(Br)=C1
1
–
1
of
1
results
1
3'-Bromo-4'-fluoroacetophenone, 98+%, Thermo Scientific Chemicals
CAS: 1007-15-4 Molecular Formula: C8H6BrFO Molecular Weight (g/mol): 217.04 MDL Number: MFCD00042466 InChI Key: SZDWTGAORQQQGY-UHFFFAOYSA-N Synonym: 3'-bromo-4'-fluoroacetophenone,3-bromo-4-fluoroacetophenone,1-3-bromo-4-fluorophenyl ethanone,1-3-bromo-4-fluorophenyl ethan-1-one,ethanone, 1-3-bromo-4-fluorophenyl,1-3-bromo-4-fluoro-phenyl-ethanone,1-acetyl-3-bromo-4-fluorobenzene,pubchem4100,acmc-1bol8,ksc499c8t PubChem CID: 70508 IUPAC Name: 1-(3-bromo-4-fluorophenyl)ethanone SMILES: CC(=O)C1=CC=C(F)C(Br)=C1