man

Test

CAS RN 1009-67-2

H3CHOO

CAS RN 1009-67-2

IUPAC Name: 2-methyl-3-phenylpropanoic acid
Synonyms: 2-methyl-3-phenylpropanoic acid
Molecular Weight (g/mol): 164.2
Molecular Formula: C10H12O2
InChi Key: MCIIDRLDHRQKPH-UHFFFAOYNA-N
SMILES: CC(CC1=CC=CC=C1)C(O)=O
Boiling Point 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)