accessibility menu, dialog, popup

Test

CAS RN 10099-66-8

ClClClLu

CAS RN 10099-66-8

IUPAC Name: lutetium(3+) trichloride
Synonyms: lutetium(III) chloride
Molecular Weight (g/mol): 281.32
Molecular Formula: Cl3Lu
InChi Key: AEDROEGYZIARPU-UHFFFAOYSA-K
SMILES: [Cl-].[Cl-].[Cl-].[Lu+3]
Quantity 
  • (3)
  • (3)
Percent Purity 
  • (2)
  • (1)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (4)
  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
  • (4)
  • (2)