accessibility menu, dialog, popup

Test

CAS RN 101342-45-4

H3COOClOHOHOHOHONH2NCH3H3COHHCl

CAS RN 101342-45-4

IUPAC Name: 7-chloro-4-(dimethylamino)-1,6,10,11,12a-pentahydroxy-6-methyl-3,12-dioxo-3,4,4a,5,5a,6,12,12a-octahydrotetracene-2-carboxamide hydrochloride
Synonyms: 7-chloro-4-(dimethylamino)-1,6,10,11,12a-pentahydroxy-6-methyl-3,12-dioxo-4,4a,5,5a-tetrahydrotetracene-2-carboxamide hydrochloride
Molecular Weight (g/mol): 515.34
Molecular Formula: C22H24Cl2N2O8
InChi Key: QYAPHLRPFNSDNH-UHFFFAOYNA-N
SMILES: Cl.CN(C)C1C2CC3C(=C(O)C4=C(O)C=CC(Cl)=C4C3(C)O)C(=O)C2(O)C(O)=C(C(N)=O)C1=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)