CAS RN 1016-77-9
CAS RN 1016-77-9
IUPAC Name:
(3-bromophenyl)(phenyl)methanone
Synonyms:
3-bromophenyl phenyl ketone
Molecular Weight (g/mol):
261.12
Molecular Formula:
C13H9BrO
InChi Key:
XNUMUNIJQMSNNN-UHFFFAOYSA-N
SMILES:
BrC1=CC=CC(=C1)C(=O)C1=CC=CC=C1
1
–
1
of
1
results
1
3-Bromobenzophenone, 97%, Thermo Scientific Chemicals
CAS: 1016-77-9 Molecular Formula: C13H9BrO Molecular Weight (g/mol): 261.118 MDL Number: MFCD00672015 InChI Key: XNUMUNIJQMSNNN-UHFFFAOYSA-N Synonym: 3-bromobenzophenone,3-bromophenyl phenyl methanone,methanone, 3-bromophenyl phenyl,m-bromobenzophenone,3-bromophenyl phenylmethanone,3-bromophenyl-phenyl-methanone,3-bromophenyl phenyl ketone,bromobenzophenone,m,3-bromobenzoylbenzene,acmc-20an8e PubChem CID: 70548 IUPAC Name: (3-bromophenyl)-phenylmethanone SMILES: C1=CC=C(C=C1)C(=O)C2=CC(=CC=C2)Br