accessibility menu, dialog, popup

Test

CAS RN 1017777-92-2

FFFH2NClCl

CAS RN 1017777-92-2

IUPAC Name: 2,3-dichloro-6-(trifluoromethyl)aniline
Synonyms: 2,3-dichloro-6-(trifluoromethyl)aniline
Molecular Weight (g/mol): 230.01
Molecular Formula: C7H4Cl2F3N
InChi Key: NESDKBLXQJESKV-UHFFFAOYSA-N
SMILES: NC1=C(C=CC(Cl)=C1Cl)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results