CAS RN 1017778-01-6
CAS RN 1017778-01-6
IUPAC Name:
1-[2-methyl-3-(trifluoromethyl)phenyl]ethan-1-one
Synonyms:
1-[2-methyl-3-(trifluoromethyl)phenyl]ethanone
Molecular Weight (g/mol):
202.18
Molecular Formula:
C10H9F3O
InChi Key:
CTVIRJUXTNXDAA-UHFFFAOYSA-N
SMILES:
CC(=O)C1=C(C)C(=CC=C1)C(F)(F)F
1
–
1
of
1
results
1
2'-Methyl-3'-(trifluoromethyl)acetophenone, 97%, Thermo Scientific™
CAS: 1017778-01-6 Molecular Formula: C10H9F3O Molecular Weight (g/mol): 202.176 MDL Number: MFCD09832283 InChI Key: CTVIRJUXTNXDAA-UHFFFAOYSA-N Synonym: 2'-methyl-3'-trifluoromethyl acetophenone,1-2-methyl-3-trifluoromethyl phenyl ethanone,1-acetyl-2-methyl-3-trifluoromethyl benzene PubChem CID: 46737515 IUPAC Name: 1-[2-methyl-3-(trifluoromethyl)phenyl]ethanone SMILES: CC1=C(C=CC=C1C(F)(F)F)C(=O)C