accessibility menu, dialog, popup

Test

CAS RN 10294-40-3

CAS RN 10294-40-3

IUPAC Name: barium(2+) dioxochromiumbis(olate)
Synonyms: barium chromate
Molecular Weight (g/mol): 253.32
Molecular Formula: BaCrO4
InChi Key: QFFVPLLCYGOFPU-UHFFFAOYSA-N
SMILES: [Ba++].[O-][Cr]([O-])(=O)=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (2)
Percent Purity 
  • (2)
1x1 image pixel for data collection

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.