CAS RN 1214336-88-5
CAS RN 1214336-88-5
IUPAC Name:
methyl 6-bromo-5-fluoropyridine-3-carboxylate
Synonyms:
methyl 6-bromo-5-fluoropyridine-3-carboxylate
Molecular Weight (g/mol):
234.02
Molecular Formula:
C7H5BrFNO2
InChi Key:
PXHKAVXJQLVUBI-UHFFFAOYSA-N
SMILES:
COC(=O)C1=CC(F)=C(Br)N=C1
1
–
1
of
1
results
1
Methyl 6-bromo-5-fluoronicotinate, 98%, Thermo Scientific Chemicals
CAS: 1214336-88-5 Molecular Formula: C7H5BrFNO2 Molecular Weight (g/mol): 234.02 MDL Number: MFCD14698110 InChI Key: PXHKAVXJQLVUBI-UHFFFAOYSA-N Synonym: methyl 6-bromo-5-fluoronicotinate,6-bromo-5-fluoro-nicotinic acid methyl ester,2-bromo-3-fluoro-5-methoxycarbonyl pyridine,methyl6-bromo-5-fluoronicotinate,abbypharma ap-30-7441,6-bromo-5-fluoropyridine-3-carboxylate,methyl 2-bromo-3-fluoro-5-pyridinecarboxylate,3-pyridinecarboxylic acid, 6-bromo-5-fluoro-, methyl ester PubChem CID: 46311203 IUPAC Name: methyl 6-bromo-5-fluoropyridine-3-carboxylate SMILES: COC(=O)C1=CC(F)=C(Br)N=C1