accessibility menu, dialog, popup

UserName

CAS RN 1315-11-3

ZnTe

CAS RN 1315-11-3

IUPAC Name: tellanylidenezinc
Synonyms: zinc telluride
Molecular Weight (g/mol): 192.98
Molecular Formula: TeZn
InChi Key: NSRBDSZKIKAZHT-UHFFFAOYSA-N
SMILES: [Zn]=[Te]
Physical Form 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)