CAS RN 132112-35-7
CAS RN 132112-35-7
IUPAC Name:
(2S)-N-(2,6-dimethylphenyl)-1-propylpiperidine-2-carboxamide hydrate hydrochloride
Synonyms:
ropivacaine hydrate hydrochloride
Molecular Weight (g/mol):
328.88
Molecular Formula:
C17H29ClN2O2
InChi Key:
VSHFRHVKMYGBJL-CKUXDGONSA-N
SMILES:
O.Cl.CCCN1CCCC[C@H]1C(=O)NC1=C(C)C=CC=C1C
1
–
2
of
2
results
1
Ropivacaine Hydrochloride Monohydrate, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
Ropivacaine hydrochloride monohydrate, MedChemExpress
MedChemExpress Ropivacaine hydrochloride monohydrate is a potent sodium channel blocker and blocks impulse conduction via reversible inhibition of sodium ion influx in nerve fibrese. Ropivacaine is also an inhibitor of K2P (two-pore domain potassium channel) TREK-1 with an IC50 of 402.7 μM in COS-7 cell's membrane. Ropivacaine is widely used for regional anesthesia and neuropathic pain management in vivo.