CAS RN 1352792-74-5
CAS RN 1352792-74-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Verinurad, MedChemExpress
MedChemExpress Verinurad (RDEA3170) is a highly potent and specific URAT1 inhibitor with an IC50 of 25 nM.