man

Test

CAS RN 13734-41-3

H3COHHNCH3OOOCH3CH3CH3

CAS RN 13734-41-3

IUPAC Name: 2-{[(tert-butoxy)carbonyl]amino}-3-methylbutanoic acid
Synonyms: 2-[(tert-butoxycarbonyl)amino]-3-methylbutanoic acid
Molecular Weight (g/mol): 217.27
Molecular Formula: C10H19NO4
InChi Key: SZXBQTSZISFIAO-UHFFFAOYNA-N
SMILES: CC(C)C(NC(=O)OC(C)(C)C)C(O)=O
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)