CAS RN 1449277-10-4
CAS RN 1449277-10-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
GNE-495, MedChemExpress
MedChemExpress GNE-495 is a potent and selective MAP4K4 inhibitor with an IC50 of 3.7 nM.