CAS RN 146939-27-7
CAS RN 146939-27-7
IUPAC Name:
hydrogen 5-{2-[4-(1,2-benzothiazol-3-yl)piperazin-1-yl]ethyl}-6-chloro-2,3-dihydro-1H-indol-2-one hydrate chloride
Synonyms:
zeldox ziprasidone hydrochloride monohydrate ziprasidone hydrochloride hydrate
Molecular Weight (g/mol):
467.41
Molecular Formula:
C21H24Cl2N4O2S
InChi Key:
ZCBZSCBNOOIHFP-UHFFFAOYSA-N
SMILES:
[H+].O.[Cl-].ClC1=CC2=C(CC(=O)N2)C=C1CCN1CCN(CC1)C1=NSC2=CC=CC=C12
1
–
2
of
2
results
1
Ziprasidone, Lipomed
Lipomed delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose Lipomed for confidence, compliance, and analytical excellence.
2
Ziprasidone, MedChemExpress
MedChemExpress Ziprasidone (CP-88059), an orally active antipsychotic agent, is a combined 5-HT and dopamine receptor antagonist. Ziprasidone mesylate trihydrate has affinities for Rat D2 (Ki=4.8 nM), 5-HT2A (Ki=0.42 nM) and 5-HT1A (Ki=3.4 nM).