CAS RN 1613724-42-7
CAS RN 1613724-42-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
Tocris Bioscience™ HTH 01-015
Potent and selective NUAK1 inhibitor
2
HTH-01-015, MedChemExpress
MedChemExpress HTH-01-015 is a selective NUAK1/ARK5 inhibitor (IC50 is 100 nM). HTH-01-015 inhibits NUAK1 with >100-fold higher potency than NUAK2 (IC50 of >10 μM).
3