CAS RN 17650-84-9
CAS RN 17650-84-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Nicotiflorin, MedChemExpress
MedChemExpress Nicotiflorin is a flavonoid glycoside extracted from a traditional Chinese medicine Flos Carthami. Nicotiflorin shows potent antiglycation activity and neuroprotection effects.