CAS RN 19614-12-1
CAS RN 19614-12-1
IUPAC Name:
1-bromo-2-(bromomethyl)-4-methoxybenzene
Synonyms:
1-bromo-2-(bromomethyl)-4-methoxybenzene
Molecular Weight (g/mol):
279.96
Molecular Formula:
C8H8Br2O
InChi Key:
MURVUTUZSUEIGI-UHFFFAOYSA-N
SMILES:
COC1=CC(CBr)=C(Br)C=C1
1
–
2
of
2
results
1
2-Bromo-5-methoxybenzyl bromide, 97%
CAS: 19614-12-1 Molecular Formula: C8H8Br2O Molecular Weight (g/mol): 279.959 MDL Number: MFCD00078664 InChI Key: MURVUTUZSUEIGI-UHFFFAOYSA-N Synonym: 2-bromo-5-methoxybenzyl bromide,1-bromo-2-bromomethyl-4-methoxybenzene,2-bromo-5-methoxybenzylbromide,4-bromo-3-bromomethyl anisole,2-bromo-5-methoxy-benzyl bromide,benzene, 1-bromo-2-bromomethyl-4-methoxy,4-bromo-3-bromomethyl-1-methoxybenzene,pubchem18176,acmc-20a5x2,4-bromo3-bromomethylanisole PubChem CID: 3525951 IUPAC Name: 1-bromo-2-(bromomethyl)-4-methoxybenzene SMILES: COC1=CC(=C(C=C1)Br)CBr