CAS RN 2095704-43-9
CAS RN 2095704-43-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
SLV-2436, MedChemExpress
MedChemExpress SLV-2436 is a highly potent and ATP-competitive inhibitor of MNK1 and MNK2 with IC50s of 10.8 nM and 5.4 nM, respectively.