CAS RN 224169-27-1
CAS RN 224169-27-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
GLN-1062, MedChemExpress
MedChemExpress Memogain is a pro-drug of galantamine, used for the treatment for Alzheimer’s disease.