accessibility menu, dialog, popup

UserName

CAS RN 2343-22-8

HNF

CAS RN 2343-22-8

IUPAC Name: 5-fluoro-2,3-dihydro-1H-indole
Synonyms: 5-fluoro-2,3-dihydro-1H-indole
Molecular Weight (g/mol): 137.16
Molecular Formula: C8H8FN
InChi Key: NXQRMQIYCWFDGP-UHFFFAOYSA-N
SMILES: FC1=CC2=C(NCC2)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results