accessibility menu, dialog, popup

UserName

CAS RN 23853-10-3

H3COONH

CAS RN 23853-10-3

IUPAC Name: 1-(2-ethoxy-2-oxoethyl)piperidin-1-ium
Synonyms: 1-(2-ethoxy-2-oxoethyl)piperidin-1-ium
Molecular Weight (g/mol): 172.25
Molecular Formula: C9H18NO2
InChi Key: LXZHLNWWDDNIOJ-UHFFFAOYSA-O
SMILES: CCOC(=O)C[NH+]1CCCCC1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)