accessibility menu, dialog, popup

UserName

CAS RN 26961-27-3

H3CONNH2OCH3

CAS RN 26961-27-3

IUPAC Name: 2-amino-4,5-dimethoxybenzonitrile
Synonyms: 2-amino-4,5-dimethoxybenzonitrile
Molecular Weight (g/mol): 178.19
Molecular Formula: C9H10N2O2
InChi Key: BJAYMNUBIULRMF-UHFFFAOYSA-N
SMILES: COC1=C(OC)C=C(C#N)C(N)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results