CAS RN 2778-96-3
CAS RN 2778-96-3
IUPAC Name:
octadecyl octadecanoate
Synonyms:
stearyl stearate
Molecular Weight (g/mol):
536.97
Molecular Formula:
C36H72O2
InChi Key:
NKBWPOSQERPBFI-UHFFFAOYSA-N
SMILES:
CCCCCCCCCCCCCCCCCCOC(=O)CCCCCCCCCCCCCCCCC
1
–
3
of
3
results
1
Stearyl stearate, 98%
CAS: 2778-96-3 Molecular Formula: C36H72O2 Molecular Weight (g/mol): 536.97 MDL Number: MFCD00056225 InChI Key: NKBWPOSQERPBFI-UHFFFAOYSA-N Synonym: stearyl stearate,octadecyl stearate,octadecanoic acid, octadecyl ester,stearic acid stearyl ester,unii-5wx2egd0dk,stearic acid, stearyl ester,octadecanyl octadecanoate,5wx2egd0dk,stearic acid, octadecyl ester,cyclochem ss PubChem CID: 17720 IUPAC Name: octadecyl octadecanoate SMILES: CCCCCCCCCCCCCCCCCCOC(=O)CCCCCCCCCCCCCCCCC