accessibility menu, dialog, popup

UserName

CAS RN 28466-26-4

NH2NHN

CAS RN 28466-26-4

IUPAC Name: 1H-pyrazol-4-amine
Synonyms: 4-aminopyrazole
Molecular Weight (g/mol): 83.09
Molecular Formula: C3H5N3
InChi Key: AXINVSXSGNSVLV-UHFFFAOYSA-N
SMILES: NC1=CNN=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results