accessibility menu, dialog, popup

UserName

CAS RN 2899-28-7

NH(S)NH2OOHClCH3

CAS RN 2899-28-7

IUPAC Name: hydrogen ethyl 2-amino-3-(1H-indol-3-yl)propanoate chloride
Synonyms: hydrogen ethyl 2-amino-3-(1H-indol-3-yl)propanoate chloride
Molecular Weight (g/mol): 268.74
Molecular Formula: C13H17ClN2O2
InChi Key: PESYCVVSLYSXAK-UHFFFAOYNA-N
SMILES: [H+].[Cl-].CCOC(=O)C(N)CC1=CNC2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results