accessibility menu, dialog, popup

Test

CAS RN 3663-20-5

H3CNHCH3O

CAS RN 3663-20-5

IUPAC Name: N-(4-butylphenyl)acetamide
Synonyms: 4-butylacetanilide
Molecular Weight (g/mol): 191.27
Molecular Formula: C12H17NO
InChi Key: GDQKURDANTWHBA-UHFFFAOYSA-N
SMILES: CCCCC1=CC=C(NC(C)=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results