CAS RN 3744-87-4
CAS RN 3744-87-4
IUPAC Name:
2-{[(tert-butoxy)carbonyl]amino}propanoic acid
Synonyms:
2-[(tert-butoxycarbonyl)amino]propanoic acid
Molecular Weight (g/mol):
189.21
Molecular Formula:
C8H15NO4
InChi Key:
QVHJQCGUWFKTSE-UHFFFAOYNA-N
SMILES:
CC(NC(=O)OC(C)(C)C)C(O)=O
1
–
1
of
1
results
1
N-Boc-DL-alanine, 97%
CAS: 3744-87-4 Molecular Formula: C8H15NO4 Molecular Weight (g/mol): 189.211 MDL Number: MFCD00063124 InChI Key: QVHJQCGUWFKTSE-UHFFFAOYSA-N Synonym: boc-dl-alanine,boc-dl-ala-oh,2-tert-butoxycarbonyl amino propanoic acid,n-boc-dl-alanine,2-n-boc-amino-propionic acid,n-tert-butoxycarbonyl alanine,2-tert-butoxycarbonylamino-propionic acid,2-tert-butoxycarbonylamino propanoic acid,2-tert-butoxy carbonyl amino propanoic acid PubChem CID: 268471 IUPAC Name: 2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid SMILES: CC(C(=O)O)NC(=O)OC(C)(C)C