accessibility menu, dialog, popup

UserName

CAS RN 432001-69-9

NNOBrOOOHSS(E/Z)

CAS RN 432001-69-9

IUPAC Name: 2-[4-bromo-2-({4-oxo-3-[(pyridin-3-yl)methyl]-2-sulfanylidene-1,3-thiazolidin-5-ylidene}methyl)phenoxy]acetic acid
Synonyms: 4-bromo-2-{[4-oxo-3-(pyridin-3-ylmethyl)-2-sulfanylidene-1,3-thiazolidin-5-ylidene]methyl}phenoxyacetic acid
Molecular Weight (g/mol): 465.34
Molecular Formula: C18H13BrN2O4S2
InChi Key: IYCJJVVXEHZJHE-UHFFFAOYSA-N
SMILES: OC(=O)COC1=C(C=C2SC(=S)N(CC3=CN=CC=C3)C2=O)C=C(Br)C=C1
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results