accessibility menu, dialog, popup

UserName

CAS RN 500912-16-3

CH3FOOH

CAS RN 500912-16-3

IUPAC Name: 2-(3-fluoro-2-methylphenyl)acetic acid
Synonyms: (3-fluoro-2-methylphenyl)acetic acid
Molecular Weight (g/mol): 168.17
Molecular Formula: C9H9FO2
InChi Key: ZESQUNYKCICBQB-UHFFFAOYSA-N
SMILES: CC1=C(CC(O)=O)C=CC=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results