CAS RN 5058-13-9
CAS RN 5058-13-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Columbianadin, MedChemExpress
MedChemExpress Columbianadin, a natural coumarin from, is known to have various biological activities including anti-inflammatory and anti-cancer effects.