accessibility menu, dialog, popup

UserName

CAS RN 515-64-0

CH3NHSOONH2NH3CN

CAS RN 515-64-0

IUPAC Name: 4-amino-N-(2,6-dimethylpyrimidin-4-yl)benzene-1-sulfonamide
Synonyms: domian sulfaisodimidinum elkosine elkosil elkosin sulfisomidine (substance) sulphisomidine optima brand of sulfisomidine sodium solfisomidina erycon
Molecular Weight (g/mol): 278.33
Molecular Formula: C12H14N4O2S
InChi Key: YZMCKZRAOLZXAZ-UHFFFAOYSA-N
SMILES: CC1=CC(NS(=O)(=O)C2=CC=C(N)C=C2)=NC(C)=N1
Certifications/Compliance 
  • (2)
  • (1)
Color 
  • (1)
Concentration 
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results