CAS RN 51649-83-3
CAS RN 51649-83-3
IUPAC Name:
6-amino-3',6'-dihydroxy-3H-spiro[2-benzofuran-1,9'-xanthen]-3-one
Synonyms:
6-amino-3',6'-dihydroxyspiro[2-benzofuran-1,9'-xanthen]-3-one
Molecular Weight (g/mol):
347.33
Molecular Formula:
C20H13NO5
InChi Key:
YOAWSYSKQHLFPM-UHFFFAOYSA-N
SMILES:
NC1=CC=C2C(=O)OC3(C2=C1)C1=CC=C(O)C=C1OC1=CC(O)=CC=C31
1
–
2
of
2
results
1
MedChemExpress 6-Aminofluorescein
MedChemExpress 6-Aminofluorescein (6-AF) is a new fluorescence marker.