accessibility menu, dialog, popup

UserName

CAS RN 5216-25-1

ClClClCl

CAS RN 5216-25-1

IUPAC Name: 1-chloro-4-(trichloromethyl)benzene
Synonyms: 4-chlorobenzotrichloride
Molecular Weight (g/mol): 229.91
Molecular Formula: C7H4Cl4
InChi Key: LVZPKYYPPLUECL-UHFFFAOYSA-N
SMILES: ClC1=CC=C(C=C1)C(Cl)(Cl)Cl
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results