CAS RN 5255-17-4
CAS RN 5255-17-4
IUPAC Name:
(4R)-4-[(1R,3aS,3bS,7S,9aR,9bS,11aR)-7-hydroxy-9a,11a-dimethyl-1H,2H,3H,3aH,3bH,4H,6H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-yl]pentanoic acid
Synonyms:
cholenic acid 3β-hydroxy-5-cholenic acid D5-cholenate 3b-hydroxy-5-cholenoate 3b-hydroxychol-5-enoate 3β-hydroxy-5-cholenoic acid 3b-hydroxycholenoate 3b-hydroxy-5-cholenoic acid 3b-hydroxychol-5-enoic acid 3b-hydroxycholenoic acid
Molecular Weight (g/mol):
374.57
Molecular Formula:
C24H38O3
InChi Key:
HIAJCGFYHIANNA-QIZZZRFXSA-N
SMILES:
C[C@H](CCC(O)=O)[C@H]1CC[C@H]2[C@@H]3CC=C4C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C
1
–
1
of
1
results
1
3b-Hydroxy-5-cholenoic acid, MedChemExpress
MedChemExpress 3b-Hydroxy-5-cholenoic acid is a monohydroxy bile acid of endogenous origin and could be found in children with the syndrome of hepatic ductular hypoplasia.