CAS RN 528-21-2
CAS RN 528-21-2
IUPAC Name:
1-(2,3,4-trihydroxyphenyl)ethan-1-one
Synonyms:
gallacetophenone
Molecular Weight (g/mol):
168.15
Molecular Formula:
C8H8O4
InChi Key:
XIROXSOOOAZHLL-UHFFFAOYSA-N
SMILES:
CC(=O)C1=CC=C(O)C(O)=C1O
1
–
1
of
1
results
1
2',3',4'-Trihydroxyacetophenone, 98%
CAS: 528-21-2 Molecular Formula: C8H8O4 Molecular Weight (g/mol): 168.148 MDL Number: MFCD00002193 InChI Key: XIROXSOOOAZHLL-UHFFFAOYSA-N Synonym: gallacetophenone,1-2,3,4-trihydroxyphenyl ethanone,2',3',4'-trihydroxyacetophenone,galloacetophenone,alizarin yellow c,alizarine yellow c,2,3,4-trihydroxyacetophenone,ethanone, 1-2,3,4-trihydroxyphenyl,4-acetylpyrogallol,acetophenone, 2',3',4'-trihydroxy PubChem CID: 10706 IUPAC Name: 1-(2,3,4-trihydroxyphenyl)ethanone SMILES: CC(=O)C1=C(C(=C(C=C1)O)O)O