accessibility menu, dialog, popup

UserName

CAS RN 5356-83-2

CH3OSiCH3H3CH2C

CAS RN 5356-83-2

IUPAC Name: ethenyl(ethoxy)dimethylsilane
Synonyms: silane, ethenylethoxydimethyl-
Molecular Weight (g/mol): 130.26
Molecular Formula: C6H14OSi
InChi Key: JEWCZPTVOYXPGG-UHFFFAOYSA-N
SMILES: CCO[Si](C)(C)C=C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results