accessibility menu, dialog, popup

UserName

CAS RN 56675-37-7

CH3H3CCH3OONH(S)SHOO

CAS RN 56675-37-7

IUPAC Name: (2S)-2-{[(tert-butoxy)carbonyl]amino}-3-(thiophen-2-yl)propanoate
Synonyms: (2S)-2-[(tert-butoxycarbonyl)amino]-3-(thiophen-2-yl)propanoate
Molecular Weight (g/mol): 270.32
Molecular Formula: C12H16NO4S
InChi Key: OJLISTAWQHSIHL-VIFPVBQESA-M
SMILES: CC(C)(C)OC(=O)N[C@@H](CC1=CC=CS1)C([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)