accessibility menu, dialog, popup

UserName

CAS RN 56990-02-4

BrBrO

CAS RN 56990-02-4

IUPAC Name: 3,5-dibromobenzaldehyde
Synonyms: 3,5-dibromobenzaldehyde
Molecular Weight (g/mol): 263.92
Molecular Formula: C7H4Br2O
InChi Key: ZLDMZIXUGCGKMB-UHFFFAOYSA-N
SMILES: BrC1=CC(C=O)=CC(Br)=C1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)