accessibility menu, dialog, popup

UserName

CAS RN 57-37-4

HClH3CNCH3OOHO

CAS RN 57-37-4

IUPAC Name: hydrogen 2-(diethylamino)ethyl 2-hydroxy-2,2-diphenylacetate chloride
Synonyms: hydrogen benactyzine chloride
Molecular Weight (g/mol): 363.88
Molecular Formula: C20H26ClNO3
InChi Key: ZCEHOOLYWQBGQO-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CCN(CC)CCOC(=O)C(O)(C1=CC=CC=C1)C1=CC=CC=C1
Certifications/Compliance 
  • (1)
  • (1)
Grade 
  • (1)
Industry Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results