accessibility menu, dialog, popup

UserName

CAS RN 574-25-4

OH(R)O(R)(R)OH(S)OHNNSNNH

CAS RN 574-25-4

IUPAC Name: 9-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-6,9-dihydro-3H-purine-6-thione
Synonyms: 9-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-3H-purine-6-thione
Molecular Weight (g/mol): 284.29
Molecular Formula: C10H12N4O4S
InChi Key: NKGPJODWTZCHGF-KQYNXXCUSA-N
SMILES: OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)N1C=NC2=C1NC=NC2=S
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results