CAS RN 6019-39-2
CAS RN 6019-39-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
OAC2, MedChemExpress
MedChemExpress OAC2 is an Oct4-activating compound which activates expression through the Oct4 gene promoter.