accessibility menu, dialog, popup

Test

CAS RN 622-86-6

ClO

CAS RN 622-86-6

IUPAC Name: (2-chloroethoxy)benzene
Synonyms: benzene, (2-chloroethoxy)-
Molecular Weight (g/mol): 156.61
Molecular Formula: C8H9ClO
InChi Key: VQUYNUJARXBNPK-UHFFFAOYSA-N
SMILES: ClCCOC1=CC=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results