CAS RN 64664-99-9
CAS RN 64664-99-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MedChemExpress CUG
MedChemExpress CUG (3-Carboxyumbelliferyl-β-D-galactopyranoside) is a fluorogenic substrate (λex=386, λem=445 nm, ε=32K).