accessibility menu, dialog, popup

UserName

CAS RN 6506-37-2

ONONNNO

CAS RN 6506-37-2

IUPAC Name: 4-[2-(5-nitro-1H-imidazol-1-yl)ethyl]morpholine
Synonyms: esclama
Molecular Weight (g/mol): 226.24
Molecular Formula: C9H14N4O3
InChi Key: MDJFHRLTPRPZLY-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CN=CN1CCN1CCOCC1
Grade 
  • (1)
  • (5)
Molecular Weight (g/mol) 
  • (5)
  • (1)
Color 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (5)
  • (5)
  • (1)
  • (5)
  • (5)
  • (1)
  • (5)
  • (5)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)